Cytochalasin B ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 7(S),20(R)-Dihydroxy-16(R)-methyl-10-phenyl-24-oxa[14]cytochalasa-6(12),13(E),21(E)-triene-1,23-dione

BMLT1080005
ENZOBMLT1080005EA 170 EUR
ENZOBMLT1080005
Cytochalasin B ≥98%
Cytochalasin B

Ultra-pure; inhibits actin polymerization


Cytochalasin B is a cell-permeable fungal toxin which binds to the barbed end of actin filaments inhibiting both the association and dissociation of subunits. It causes the disruption of actin filaments and inhibition of actin polymerization. It also inhibits monosaccharide transport across cell membranes. inhibits glucose transport and platelet aggregation. Blocks adenosine-induced apoptotic body formation without affecting activation of endogenous ADP-ribosylation in leukemia HL-60 cells.


Isolated from Drechslera dematoidea

Formel: C₂₉H₃₇NO₅
Molecular Weight: 479.62 g/mol
MDL: MFCD00077704
CAS-Nummer: 14930-96-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ad0e69c0e38034cde8d241e9302eababa962577&vid=e51c21f048caecb6aa373b2c87189885e4b9a704&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR