TN-16 ≥97% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

BMLT1200050
ENZOBMLT1200050EA 165 EUR
ENZOBMLT1200050
TN-16 ≥97% (durch HPLC)
TN-16

A novel synthetic inhibitor of microtubule formation which binds to the colchicine site. It prevents the stabilization of microtubules in taxol-treated cells. Blocks the cell cycle in M-phase.


  • Microtubule inhibitor


A novel synthetic inhibitor of microtubule formation which binds to the colchicine site. It prevents the stabilization of microtubules in taxol-treated cells. Blocks the cell cycle in M-phase.

Lagertemperatur: Raumtemperatur
CAS-Nummer: 33016-12-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ad66d709644f2e6623c25e0ae0145b77d320b35&vid=097985453f0fa8afcac3f4b46047de382857de07&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR