Ansamitocin P-3 93 HPLC_ASSAY_METHOD

Lieferant: ENZO LIFE SCIENCES

Synonyme: 3-O-de[2-(acetylmethylamino)-1-oxopropyl]-3-O-(1-oxobutyl)-maytansine, Maytansinol isobutyrate, Antibiotic C 15003P3, (14S,16S,32S,33S,2R,4S,10E,12E,14R)-86-Cchloro-14-hydroxy-85,14-dimethoxy-33,2,7,10-tetramethyl-12,6-dioxo-7-aza-1(6,4)-oxazinana-3(2,3)-oxirana-8(1,3)-benzenacyclotetradecaphane-10,12-dien-4-yl isobutyrate, NSC292222

BMLT1210001
ENZOBMLT1210001EA 467 EUR
ENZOBMLT1210001
Ansamitocin P-3 93 HPLC_ASSAY_METHOD
Ansamitocin P-3

Ansamitocin P-3 is a maytansine analog which displays potent cytotoxicity against the human solid tumor cell lines A-549 and HT-29. It shares the same binding site as rhizoxin and phomopsin A on porcine brain tubulin. Maytansine and its analogs cause extensive disassembly of the microtubule and totally prevent tubulin spiralization.


  • Microtubule inhibitor


White to off-white crystalline solid.


Soluble in DMSO.

Formel: C₃₂H₄₃ClN₂O₉
Molecular Weight: 635.14 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00274586
CAS-Nummer: 66547-09-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b3d7bf22d8c671190277b991903650244e9864f&vid=244b5e373396c635c179ca14cef7f3aa85709b7c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR