SUMO-1 (93-97)-AMCA
Lieferant: ENZO LIFE SCIENCES
BMLUW05001000
ENZOBMLUW05001000EA
164
EUR
ENZOBMLUW05001000
SUMO-1 (93-97)-AMCA
Proteine und Peptide
SUMO1-AMCA is a synthetic AMCA-peptide substrate based on the C-terminal residues of SUMO1 and is a useful fluorogenic substrate for deSUMOylating enzymes, including SENP1 (BML-UW9760).
- Provides an alternative substrate to SUMO1-AMC
It has particular utility for the study of deSUMOylating activity where continuous monitoring of activity is essential.