Di-Ubiqutin (K63 linked)
Lieferant: ENZO LIFE SCIENCES
BMLUW07300050
ENZOBMLUW07300050EA 368 EUR
ENZOBMLUW07300050
Di-Ubiqutin (K63 linked)
Proteine und Peptide
Protein components produced in E. coli. Synthesised enzymatically in vitro.
Polyubiquitin chains, linked through specific lysine residues, are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of polyubiquitin modified proteins with by ubiquitin binding domain (UBDs) containing proteins.