Poly-Ubiquitin chains (linear)
Lieferant: ENZO LIFE SCIENCES
BMLUW08250001
ENZOBMLUW08250001EA 482 EUR
ENZOBMLUW08250001
Poly-Ubiquitin chains (linear)
Proteine und Peptide
For use in deubiquitinylating enzyme assays and linear polyubiquitin binding studies.
Sampler pack for use in deubiquitinylating enzyme assays and linear polyubiquitin binding studies.
Bestellinformation: Contents: Name Prod. No. Amount Mol. Weight Di-ubiquitin (Ub2) (linear) BML-UW0775 25µg 17,1 kDa Tri-ubiquitin (Ub3) (linear) BML-UW0780 25 µg 25,7 kDa Tetra-ubiquitin (Ub4) (linear) BML-UW0785 25 µg 34,2 kDa Penta-ubiquitin (Ub5) (linear) BML-UW0790 25 µg 42,8 kDa Hexa-ubiquitin (Ub6) (linear) BML-UW0795 25 µg 51,3 kDa Hepta-ubiquitin (Ub7) (linear) BML-UW0800 25 µg 59,8 kDa Octa-ubiquitin (Ub8) (linear) BML-UW0805 25µg 68.4kDa Nona-ubiquitin (Ub9) (linear) BML-UW0810 25 µg 76,9kDa Deca-ubiquitin (Ub10) (linear) BML-UW0815 25 µg 86,5 kDa Undeca-ubiquitin (Ub11) (linear) BML-UW0820 25 µg 94,0 kDa