Poly-Ubiquitin chains (linear)
Lieferant: ENZO LIFE SCIENCES
For use in deubiquitinylating enzyme assays and linear polyubiquitin binding studies.
Sampler pack for use in deubiquitinylating enzyme assays and linear polyubiquitin binding studies.
Bestellinformation: Contents: Name Prod. No. Amount Mol. Weight Di-ubiquitin (Ub2) (linear) BML-UW0775 25µg 17,1 kDa Tri-ubiquitin (Ub3) (linear) BML-UW0780 25 µg 25,7 kDa Tetra-ubiquitin (Ub4) (linear) BML-UW0785 25 µg 34,2 kDa Penta-ubiquitin (Ub5) (linear) BML-UW0790 25 µg 42,8 kDa Hexa-ubiquitin (Ub6) (linear) BML-UW0795 25 µg 51,3 kDa Hepta-ubiquitin (Ub7) (linear) BML-UW0800 25 µg 59,8 kDa Octa-ubiquitin (Ub8) (linear) BML-UW0805 25µg 68.4kDa Nona-ubiquitin (Ub9) (linear) BML-UW0810 25 µg 76,9kDa Deca-ubiquitin (Ub10) (linear) BML-UW0815 25 µg 86,5 kDa Undeca-ubiquitin (Ub11) (linear) BML-UW0820 25 µg 94,0 kDa