Human recombinant pro-ISG15 (from E. coli), HIS Tag
Lieferant: ENZO LIFE SCIENCES
BMLUW92300500
ENZOBMLUW92300500EA
207
EUR
ENZOBMLUW92300500
Human recombinant pro-ISG15 (from E. coli), HIS Tag
Proteine und Peptide
Produced in E. coli. Human pro-ISG15 is fused at the N-terminus to a His-tag.
ISG15 is expressed in its pro-form with a short C-terminal extension, which is processed to expose the C-terminal glycine residue that is essential for conjugation to target proteins.