Gallocyanin ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLWN1180500
ENZOBMLWN1180500EA 73.7 EUR
ENZOBMLWN1180500
Gallocyanin ≥98% (durch TLC)
Gallocyanine

DKK1 inhibitor


Inhibits DKK1 of the Wnt pathway.

Formel: C₁₅H₁₃ClN₂O₅
Molecular Weight: 336,73 g/mol
MDL: MFCD00011926
CAS-Nummer: 1562-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR