Z-Pro-Nle-Asp-CHO
Lieferant: ENZO LIFE SCIENCES
BMLZW94900100
ENZOBMLZW94900100EA
149
EUR
ENZOBMLZW94900100
Z-Pro-Nle-Asp-CHO
Proteine und Peptide
Highly specific aldehyde based inhibitor for the caspase-like peptidic activity of the 20S proteasome.