NGAL (Human) ELISA kit
Lieferant: ENZO LIFE SCIENCES
A colorimetric immunometric ELISA kit for the measurement of NGAL in human samples. Absorbance is read at 450 nm.
- Fast results from up to 40 samples in duplicate in just 4 hours per 96 well plate
- Highly sensitive measurement, detecting as low as 4 pg/ml of NGAL
- Ready-to-use pre-coated plate and individual calibrators save time and reduce errors
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of NGAL in human urine, plasma, serum, tissue extracts, and culture supernatants.
Lieferumfang: Kit consists of 12 strips of 8 microwells each coated with anti-NGAL (96 wells) + frame, sample diluent, NGAL calibrators 1-8; 0, 10, 25, 50, 100, 250, 500, 1000 pg/ml, 25X wash solution concentrate, biotinylated NGAL antibody, HRP-Streptavidin conjugate, TMB substrate and stop solution.