Proteinase K
Lieferant: ENZO LIFE SCIENCES
ENZ33801
ENZOENZ33801EA 88 EUR
ENZOENZ33801
Proteinase K
Enzyme
ENZO Proteinase K is specifically designed for use in the pretreatment of formalin-fixed tissue and paraformaldehyde-fixed cells in preparation for in situ hybridization and detection studies.