BIO-PROBE® Lambda probe
Lieferant: ENZO LIFE SCIENCES
Negative control probe for in situ hybridization.
The Lambda BIO-PROBE® labeled probe is prepared by nick translation of the lambda bacteriophage strain c1857ind1 Sam7. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).