BK virus BioProbe®
Lieferant: ENZO LIFE SCIENCES
ENZ40848
ENZOENZ40848EA
420
EUR
ENZOENZ40848
BK virus BioProbe®
Nukleinsäure-Reagenzien
Primer und Proben
The BK Virus BioProbe® labeled probe is prepared by nick translation of a clone of the entire BK virus genome. BK virus, a simian polyoma virus, is related to SV40 and also bears some similarity to the human virus JC virus. BK virus is highly tumorigenic in hamsters. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).