Blur 8 BioProbe®
Lieferant: ENZO LIFE SCIENCES
ENZ40849
ENZOENZ40849EA
420
EUR
ENZOENZ40849
Blur 8 BioProbe®
Nukleinsäure-Reagenzien
Primer und Proben
The Blur 8 BioProbe® labeled probe is prepared by nick translation of a clone of the human alu repeat sequence. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).