Bio-17-ATP

Lieferant: ENZO LIFE SCIENCES

ENZ42817
ENZOENZ42817EA 379 EUR
ENZOENZ42817
Bio-17-ATP
Nukleinsäure-Reagenzien Nukleotide

Bio-17-ATP (Biotin-17-adenosine-5’-triphosphate) can replace ATP for in vitro transcription reaction catalysed by T3, T7 or SP6 RNA polymerases.


  • Appearance: Clear, colorless liquid
  • Purity: ≥93% (HPLC)
  • Concentration: 10 mM
  • Extinction coefficient: 18000 M⁻¹ cm⁻¹ (267 nm, pH 7,0)


The biotin-labelled RNA transcripts produced by these reactions are suitable for a wide range of applications such as nucleic acid hybridisation, sequencing, and genome analysis.


The biotin-labelled hybridised probes can be detected by a reporter molecule linked to streptavidin, avidin, or anit-biotin antibody. Such a complex can be detected directly, e.g. by excitation of a fluorophore conjugated to streptavidin, or indirectly, e.g. using an enzyme conjugate that can produce an insoluble coloured precipitate.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7768c0f17e498307d4f6fb8d362afd92133cad&vid=0402b1d1202045ad5ff0489e6c9065eb486d70bb&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR