DNA isolation kit, EPIXTRACT®
Lieferant: ENZO LIFE SCIENCES
ENZ450180050
ENZ450180100
ENZOENZ450180050EA
247
EUR
ENZOENZ450180050
ENZOENZ450180100
DNA isolation kit, EPIXTRACT®
Nukleinsäure-Reagenzien
Reinigungskits und Reagenzien für Nukleinsäure
EPIXTRACT® DNA isolation kit provides a simple method to isolate DNA from plasma, serum, and body fluids.
- Rapid, provide results in less than 15 minutes
- Safe to use, contains only non-toxic reagents and no phenol chloroform
- Highly efficient, isolate DNA from plasma or serum containing as low as 0,1 ng DNA
Lieferumfang: Supplied with digestion solution, digestion powder, DNA isolation buffer, DNA elution solution, spin column and collection tube.