Red detection kit (GFP-CERTIFIED®), LYSO-ID®
Lieferant: ENZO LIFE SCIENCES
LYSO-ID® Red Detection Kit (GFP-CERTIFIED®) is an acidic organelle-selective dye for live cell staining of lysosomes. It is an ideal choice for fluorescence co-localisation imaging with GFP-tagged proteins and providing spectrally pure red signal.
- No photo conversion to a green emitting dye
- No complicating metachromatic dual emission artifacts, facilitating analysis with GFP, FITC and other fluorescent probes
- Highly resistant to photobleaching and concentration quenching
- Ensures strong and consistent fluorescence signal, even after extended viewing periods
LYSO-ID® Red Detection Kit is for use with GFP-expressing cell lines, as well as cells expressing blue, cyan or yellow fluorescent proteins.
Lieferumfang: Kit contains LYSO-ID® Red Detection Reagent, Hoechst 33342 nuclear stain, chloroquine control and 10X assay buffer.