Membrane potential detection kit, MITO-ID®
Lieferant: ENZO LIFE SCIENCES
ENZ510180025 ENZ51018K100
ENZOENZ510180025EA 238 EUR
ENZOENZ510180025 ENZOENZ51018K100
Membrane potential detection kit, MITO-ID®
Tests Zelltests
MITO-ID® membrane potential detection kit monitors energetic status using a simple mix-and-read, no-wash protocol. It measures mitochondrial membrane potential with a cationic dye that fluoresces either green or orange depending upon membrane potential status.
  • Highly sensitive detection of mitochondrial membrane potential decline
  • Aids in the drug research process in preclinical drug safety assessment (ADME-Tox)
  • Suitable for chemical/environmental toxicity screening and characterisation
  • True mix-and-read homogeneous assay for live cells
  • Optimized for the measurement of mitochondria membrane potential (MMP) and cell viability by fluorescence microscopy and flow cytometry
Cell-based assays for evaluating the functional status of mitochondria are useful tools for elucidating the role of mitochondrial activity in drug-induced toxicity, the apoptosis cascade and other cellular and biochemical processes.
Lieferumfang: Kit contains MITO-ID® MP detection reagent, necrosis detection reagent, CCCP control, 10X assay buffer 1 and 50X assay buffer 2.