CYTO-ID® Autophagy detection kit
Lieferant: ENZO LIFE SCIENCES
ENZ51031K200
ENZOENZ51031K200EA 620 EUR
ENZOENZ51031K200
CYTO-ID® Autophagy detection kit
Tests Zelltests
The CYTO-ID® Autophagy detection kit provides a rapid, specific and quantitative approach for monitoring autophagy in live cells by fluorescence microscopy and flow cytometry.
CYTO-ID® Autophagy Detection Kit measures autophagic vacuoles and monitors autophagic flux in live cells using a novel dye that selectively labels autophagic vacuoles. The dye has been optimized through the identification of titratable functional moieties that allow for minimal staining of lysosomes while exhibiting bright fluorescence upon incorporation into pre-autophagosomes, autophagosomes, and autolysosomes (autophagolysosomes).
Lieferumfang: Kit contains CYTO-ID® Green Detection Reagent, 50 µl, Hoechst 33342 Nuclear Stain, 50 µl Autophagy Inducer (Rapamycin, 25 nmol) and 10X Assay Buffer, 30 ml.