FLUO 3/AM ≥95% (durch HPLC), ultrarein

Lieferant: ENZO LIFE SCIENCES

Synonyme: 4-(6-Acetoxymethoxy-2,7-dichloro-3-oxo-9-xanthenyl)-4′-methyl-2,2′(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, 1-[Amino-5-(2,7-dichloro-6-acetomethoxy-3-oxo-3H-xanthen-9-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid pentaacetoxy-methyl ester

ENZ52004
ENZOENZ52004EA 355 EUR
ENZOENZ52004
FLUO 3/AM ≥95% (durch HPLC), ultrarein
FLUO 3/AM

Calcium dye


Fluo-3 AM is widely used in flow cytometry, confocal laserscanning microscopy and fluorescence spectroscopy. The probe has proved particularly valuable for determining GPCR function via cell-based high-throughput screening. Fluo-3 AM is a membrane permeable probe and can be passively loaded in cells by simple incubation. Once inside the cell, Fluo-3 becomes fluorescent in the presence of free Ca (II) . Fluo-3 is a long wavelength calcium indicator probe (488 nm excitable). The longer wavelength of absorption is convenient for minimizing photodamage in cells. Wavelength Maxima: Excitation 506 nm, Emission 526 nm

Formel: C₅₁H₅₀Cl₂N₂O₂₃
Molecular Weight: 1129.85 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 121714-22-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a1569467a333cfc5c632b8c9b6785cb411a3bee&vid=20d79d5b28cc06fe6f8e3ef0479eb6653e24899e&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR