FURA-2 pentapotassium salt ≥95% (durch HPLC), ultrarein

Lieferant: ENZO LIFE SCIENCES

Synonyme: 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid . 5K, FURA-2

ENZ52007
ENZOENZ52007EA 220 EUR
ENZOENZ52007
FURA-2 pentapotassium salt ≥95% (durch HPLC), ultrarein
FURA-2 pentapotassium salt

Calcium dye


Fura-2 is one of the most widely used calcium indicators for homogenous quantitative dual wavelength ratiometric cell measurements. Fura-2 is particularly useful for digital imaging microscopy. It is less susceptible to photobleaching than Indo-1. The probe is excited only by UV light, which results in significantly less interference by visible wavelength excitable fluorescent compounds. One application of the probe is as a useful counter screen tool for GPCR and calcium channels identified using calcium indicators excited at 488 nm in primary screens. Fura-2 is a membrane impermeable probe and can be loaded in cells by microinjection, electroporation or scrape loading. The acetomethoxy (AM) version is necessary for passive cell loading. Wavelength Maxima: Excitation 363 nm, Emission 512 nm.

Formel: C₂₉H₂₂K₅N₃O₁₄
Molecular Weight: 832 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 113694-64-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a19a04832a861db45c3d77467aaeb251177e05ab&vid=4ef2a73d683df032850b0be7a09278dc7331a713&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR