Rhod-2 AM (Bromide) ≥90% (durch HPLC), ultrarein
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Rhod-2 AM ester, 1-[2-Amino-5-(3-dimethylamino-6-dimethylammonio-9-xanthenyl)phenoxy]-2-(2-amino-5-methylphenoxy)]ethane-N,N,N',N'-tetraacetic acid, tetraacetoxy methyl ester . Br
Calcium dye
Rhod-2 AM has the longest fluorescent emission signal of the commonly used calcium indicators. It contains a rhodamine-like fluorophore, making it suitable for use with argon and krypton laser excitation sources. Rhod-2 signal intensity one of the strongest of all the calcium probes. Rhod 2 is especially suitable probe for intracellular calcium monitoring by confocal laser-scanning microscopy and flow cytometry. The dissociation constant of Rhod 2 with calcium is among the highest of all the fluorescent calcium probes, providing a wider range for monitoring calcium concentration. Rhod-2 AM is an acetoxymethyl ester derivative of Rhod-2 that can be easily loaded into cells by passive incubation. Wavelength Maxima: Excitation 549 nm, Emission 578 nm.
Formel:
C₅₂H₅₉BrN₄O₁₉ Molecular Weight: 1124,0 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
145037-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...