Coelenterazine ≥90% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
Calcium dye.
Colenterazine is useful for monitoring calcium in live cells and tissues, in reporter gene assays, for superoxide anion detection and in drug high-throughput screening applications. Coelenterazine is useful for reconstituting aequorin in cells that have been transfected with apo-aequorin cDNA. The aequorin complex comprises a 22 kDa apoaequorin protein, molecular oxygen and coelenterazine. In the presence of Ca(II), coelenterazine is oxidized to coelenteramide, yielding the release of carbon dioxide and blue light. Unlike fluorescent Ca(II) indicators, Ca²⁺-bound aequorin can be detected without illuminating the sample, thereby eliminating interference from autofluorescence. Wavelength Maxima: Excitation 429 nm, Emission 466 nm
Soluble in 100% ethanol.
Formel:
C₂₆H₂₁N₃O₃ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00467176 CAS-Nummer: 55779-48-1 |
|
Spezifikation Testergebnisse
Purity | ≥90% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...