BCECF AM ester ≥95% (durch HPLC), ultrarein

Lieferant: ENZO LIFE SCIENCES

Synonyme: BCECF AM

ENZ52102
ENZOENZ52102EA 323 EUR
ENZOENZ52102
BCECF AM ester ≥95% (durch HPLC), ultrarein
BCECF AM ester

pH indicator


BCECF AM (2’,7’-bis-(2-carboxyethyl)-5-(and-6)-carboxyfluorescein acetoxymethyl ester) is a well established fluorescent probe that enables ratiometric monitoring of cellular pH. Intracellular pH plays an important role in modulating cellular events, including growth, calcium regulation, enzymatic activity, receptor-mediated signal transduction, ion transport, endocytosis, chemotaxis, adhesion and other cellular processes. Once inside, cellular esterases cleave the AM groups yielding a more hydrophilic BCECF trapped inside the cell. Wavelength Maxima: Excitation 505 nm, Emission 520 nm.

Formel: C₄₂H₄₀O₂₁
Molecular Weight: 808.69 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00036969
CAS-Nummer: 117464-70-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8c0404448381cc691798096a804b82ee29a57d2&vid=856b228fbb3a33e05bf364252e85894be3d2ac1d&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR