5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate ≥98% (durch HPLC), ultrarein

Lieferant: ENZO LIFE SCIENCES

Synonyme: Carboxy-DCFDA, 5-(6)-Carboxydichlorofluorescein diacetate, 5(6)-CDCFDA, CDCFDA

ENZ52103
ENZOENZ52103EA 165 EUR
ENZOENZ52103
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate ≥98% (durch HPLC), ultrarein
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate

pH indicator


5(6)-CDCFDA is an amine–reactive fluorescein diacetate (FDA) derivative used to prepare a variety of FDA conjugates. 5(6)-CDCFDA is a non-fluorescent molecule that diffuse into cells and is hydrolyzed by intracellular non-specific esterases, yielding a fluorescent probe. The probe accumulates exclusively in cells with intact cell membranes. 5(6)-CFCFDA does not stain dead cells. 5(6)-CDCFDA labeling can be used for monitoring cells by flow cytometry or fluorescence microscopy. The probe is also frequently used to monitor reactive oxygen species (ROS) generation in live cells. Wavelength Maxima: Excitation 504 nm, Emission 529 nm.

Formel: C₂₅H₁₄Cl₂O₉
Molecular Weight: 529,28 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD08459585
CAS-Nummer: 127770-45-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f33dec0ead2421febd8c529b7039309bc054be27&vid=6ce1ac9ccac0d785237ae3cbc9891f8ebf34d905&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR