Zinquin ethyl ester ≥98% (durch HPLC), ultrarein fluoreszierender Indikator, für die Bestimmung von Zink

Lieferant: ENZO LIFE SCIENCES

Synonyme: 2-[[2-Methyl-8-[[(4-methylphenyl)sulphonyl]amino]-6-quinolinyl]oxy]-acetic acid ethyl ester

ENZ52151
ENZOENZ52151EA 235 EUR
ENZOENZ52151
Zinquin ethyl ester ≥98% (durch HPLC), ultrarein fluoreszierender Indikator, für die Bestimmung von Zink
Zinquin ethyl ester

Zinc dye


Zinquin ethyl ester is a lipophilic, zinc-sensitive, fluorescent derivative of Zinquin that is able to penetrate cell membranes. Cleavage of the ethyl ester group via cytosolic esterases in living cells impedes its efflux across the plasma membrane. It is selective for Zn (II) in the presence of physiological concentrations of Ca (II) and Mg (II) ions. Fluorescence of Zinquin ethyl ester is enhanced upon binding Zn (II), but the probe does not demonstrate a shift or change in emission wavelength. The probe is UV-excitable and emits in the blue reqion of the spectrum. Zinquin ethyl ester has been used to monitor intracellular zinc fluxes associated with apoptosis. Wavelength Maxima: Excitation 368nm, Emission 490nm

Formel: C₂₁H₂₂N₂O₅S
Molecular Weight: 414,47 g/mol
CAS-Nummer: 181530-09-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac72f6029f82707cd2600492d70798c9014edc34&vid=4b82444f295a2038301f0de44a4c3e0e24419427&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR