TSQ ≥98% (durch HPLC), ultrarein

Lieferant: ENZO LIFE SCIENCES

Synonyme: N-(6-Methoxy-8-quinolyl)-p-toluenesulphonamide

ENZ52153
ENZOENZ52153EA 165 EUR
ENZOENZ52153
TSQ ≥98% (durch HPLC), ultrarein
TSQ

Zinc dye


TSQ is one of the most efficient membrane-permeable fluorescent probes for zinc (II). It is selective for Zn (II) in the presence of physiological concentrations of Ca (II) and Mg (II) ions. Fluorescence of Zinquin ethyl ester is enhanced upon binding Zn (II), but the probe does not demonstrate a shift or change in emission wavelength. TSQ forms a largely insoluble 2:1 (Ligand-Metal) complex with zinc and emits blue light upon excitation at 365 nanometers. TSQ has been extensively applied to study Zn (II) in mossy fibers of the hippocampus. TSQ has been used to monitor intracellular zinc fluxes associated with apoptosis as well. Wavelength Maxima: Excitation 344 nm, Emission 385 nm.

Formel: C₁₇H₁₆N₂O₃S
Molecular Weight: 328,38 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 109628-27-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=827a89ee88048e17011c6d873fa27d55cd833ae3&vid=996ba72f08d275292d50e62fc434a76a8b461849&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR