N-(3-Triethylammoniumpropyl)-4-(6-(4-(diethylamino)phenyl)hexatrienyl)pyridinium dibromide (MM 4-64) ≥90% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

ENZ52252
ENZOENZ52252EA 421 EUR
ENZOENZ52252
N-(3-Triethylammoniumpropyl)-4-(6-(4-(diethylamino)phenyl)hexatrienyl)pyridinium dibromide (MM 4-64) ≥90% (durch HPLC)
N-(3-Triethylammoniumpropyl)-4-(6-(4-(diethylamino)phenyl)hexatrienyl)pyridinium dibromide (MM 4-64)

Neuronal and autophagy detection in yeast detection.


MM 4-64 is a cationic dye, equivalent to membrane probe FM® 4-64. MM 4-64 is a lipophilic styryl dye that is used as a vital stain to follow bulk membrane-internalization and transport to the vacuole in yeast. MM 4-64 is also a sensitive reporter of vacuolar dynamics, detecting such events as segregation structure formation during mitosis, vacuole fission/fusion events, and vacuolar morphology in different classes of vacuolar protein sorting mutants. Wavelength Maxima: Excitation 558 nm, Emission 734 nm.

Formel: C₃₀H₄₅Br₂N₃
Lagertemperatur: Gefrierschrank
MDL: MFCD00467819
CAS-Nummer: 162112-35-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=947cf1974107417ebc760ea44087b18b541d786a&vid=8f593f4364a17a23968d3d9a5a0582349ffbf498&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR