3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥95% (durch HPLC), ultrarein

Lieferant: ENZO LIFE SCIENCES

Synonyme: 3-Hexyl-2-[3-(3-hexyl-1,3-benzoxazol-2(3H)-ylidene)-1-propen-1-yl]-1,3-benzoxazol-3-ium iodide, DiOC6(3) iodide, DiOC6(3), 3,3'-Dihexyloxacarbocyanine iodide, 3-Hexyl-2-[3-(3-hexyl-2(3H)benzoxazolylidene)-1-propenyl]benzoxazolium iodide

ENZ52303
ENZOENZ52303EA 165 EUR
ENZOENZ52303
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥95% (durch HPLC), ultrarein
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide]

Mitochondria dye


DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures. The fluorescence of these environment sensitive dyes is greatly enhanced when incorporated into membranes or bound to lipophilic biomolecules such as proteins although they are weakly fluorescent in water. Once applied to cells, these dyes diffuse laterally within the cellular plasma membranes, resulting in even staining of the entire cell at their optimal concentrations. DiOC6(3) is a green fluorescent membrane dye that has been used to detect mitochondrial membrane potential in live cells. The probe has also been employed in the analysis of apoptotic pathways by multiparametric flow cytometry. The dye has also been employed with the microfluidic Agilent 2100 bioanalyzer for assessing mitochondrial membrane potential. Wavelength Maxima: Excitation 482 nm, Emission 504 nm.

Formel: C₂₉H₃₇IN₂O₂
Molecular Weight: 572,531 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00011954
CAS-Nummer: 53213-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aca288e4b8b097c3fa0a37e364f08baa0daaaf67&vid=d72ab5ec5bf680a5d1ffaae791fac820469a197c&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR