Acridine Orange 10-nonyl bromide (NAO) ≥95% (durch HPLC), ultrarein

Lieferant: ENZO LIFE SCIENCES

Synonyme: 3,6-Bis(dimethylamino)-10-nonylacridinium bromide, Nonyl acridine orange, NAO, 10-Nonylacridine orange bromide

ENZ52306
ENZOENZ52306EA 231 EUR
ENZOENZ52306
Acridine Orange 10-nonyl bromide (NAO) ≥95% (durch HPLC), ultrarein
Acridine Orange 10-nonyl bromide (NAO)

Mitochondria dye


10-N-Nonyl acridine orange (NAO) is an acridine orange derivative and is generally used as a fluorescent marker of the inner mitochondrial membrane in whole cells. It is believed to bind to negatively charged phospholipids. NAO accumulation in the cell seems to be related to specific interactions with mitochondrial membrane proteins and/or lipids, such as cardiolipin, and is largely independent of mitochondrial membrane potential. With respect to apoptosis, the presence of mitochondrial membrane potential can be probed with Rhodamine123 while the structure and integrity of mitochondria can be assessed using 10-N-nonyl-acridine orange. Wavelength Maxima: Excitation 495 nm, Emission 519 nm.

Formel: C₂₆H₃₈BrN₃
Lagertemperatur: Gefrierschrank
MDL: MFCD00467605
CAS-Nummer: 75168-11-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22fa5fb86b81969c3589d4c5c154738cebb8d6de&vid=b6126e837976f5393be1bc2887c502c633842454&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR