Tetramethylrhodamine ethyl ester perchlorate (TMRE) ≥95% (durch HPLC), CELLESTIAL®, ultrarein Mitochondrien-Färbemittel
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
Synonyme:
Tetramethylrhodamine Ethyl Ester Perchlorate, TMRE, 3,6-bis(Dimethylamino)-9-[2-(ethoxycarbonyl)phenyl]-xanthylium perchlorate
Positively charged rhodamine dyes (such as rhodamine esters and rosamines) are selectively localized in mitochondria, thus they are widely used for labeling mitochondria of live cells. Like JC-1,TMRE is widely used for measuring mitochondrial membrane potential, in addition to selectively staining mitochondria. Real-time imaging of mitochondrial membrane potential in individual cardiomyocytes within perfused rat hearts has been demonstrated with this dye, using 2-photon laser-scanning microscopy.
Wavelength Maxima: Excitation 549nm, Emission 574nm
Formel:
C₂₆H₂₇ClN₂O₇ Molecular Weight: 514,95 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00467973 CAS-Nummer: 115532-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...