bisbenzimide H-33342 trihydrochloride (Hoechst 33342) ≥95% (durch HPLC), ultrarein Farbstoff

Lieferant: ENZO LIFE SCIENCES

Synonyme: Hoechst 33342, 2′-(4-Ethoxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bi-1H-benzimidazole trihydrochloride, HOE 33342, 2'-(4-Ethoxyphenyl)-5-(4-methylpiperazin-1-yl)-1H,1'H-2,5'-bibenzo[d]imidazole, bisBenzimide

ENZ52401
ENZOENZ52401EA 190 EUR
ENZOENZ52401
bisbenzimide H-33342 trihydrochloride (Hoechst 33342) ≥95% (durch HPLC), ultrarein Farbstoff
bisbenzimide H-33342 trihydrochloride (Hoechst 33342)

DNA dye


Hoechst 33342 is fluorescent probe useful for detecting DNA by fluorescence microscopy and flow cytometry. Hoechst 33342 may be used on live or fixed cells and is also applicable for cell cycle analysis and monitoring DNA condensation by flow cytometry. An additional ethyl group renders Hoechst 33342 more lipophilic, and thus more readily cell permeable than Hoechst 33258 . Exclusion of Hoechst 33342 dye is a characteristic common to stem cells, as well as chemotherapy-resistant cancer cells. Wavelength Maxima: Excitation 350nm, Emission 461nm

Formel: C₂₇H₂₈N₆O·3HCl
Lagertemperatur: Gefrierschrank
MDL: MFCD00012679
CAS-Nummer: 23491-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4beb6fa5e65e2f3bb60723371acbfd3961cbdff&vid=43eadb5493e654016a2c0c73dfa9a9a8a65aaf25&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR