bisBenzimide H 33258 trihydrochloride (Hoechst 33258) ≥95% (durch HPLC), ultrarein Farbstoff
Lieferant: ENZO LIFE SCIENCES
Synonyme:
BBIH, HOE 33258, Hoechst 33258
ENZ52402
ENZOENZ52402EA
190
EUR
ENZOENZ52402
bisBenzimide H 33258 trihydrochloride (Hoechst 33258) ≥95% (durch HPLC), ultrarein Farbstoff
bisBenzimide H 33258 trihydrochloride (Hoechst 33258)
DNA dye
Hoechst 33258 is a fluorescent probe useful for detecting DNA by fluorescence microscopy and flow cytometry. Hoechst 33258 may be used on live or fixed cells and is also applicable for cell cycle analysis and monitoring DNA condensation by flow cytometry. Hoechst 33358 is less cell permeable that other derivatives, however this dye also allows for quantitative measurements when plotted in a standard emission-to-content curve. Wavelength Maxima: Excitation 352nm, Emission 461nm
Formel:
C₂₅H₂₄N₆O·3HCl Molecular Weight: 533,88 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00012679 CAS-Nummer: 23491-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...