4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) ≥95% (durch HPLC), CELLESTIAL®, ultrarein fluoreszierender Farbstoff

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: 2-(4-Amidinophenyl)indole-6-carboxamidine Dihydrochloride, 2-(4-Carbamimidoylphenyl)-1H-indole-6-carboximidamide dihydrochloride, DAPI.2HCl, 2-(4-Carbamimidoylphenyl)indole-6-carboximidamide Dihydrochloride, DAPI 2HCl

ENZ52404
ENZOENZ52404EA 839 EUR
ENZOENZ52404
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) ≥95% (durch HPLC), CELLESTIAL®, ultrarein fluoreszierender Farbstoff
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI)

DNA dye


DAPI is a fluorescent dye that binds nucleic acids and demonstrates a high affinity for DNA. It is used extensively in fluorescence microscopy, flow cytometry and microplate assays. DAPI is cell permeable and is readily used for detecting both live, dead and fixed cells. DAPI emits in the blue spectral range and is ideal for multiplexed assays inclusive of green-fluorescent molecules like fluorescein and green fluorescent protein (GFP), or red-fluorescent stains like Texas Red. Aside from labeling cell nuclei, DAPI is also used for the detection of mycoplasma or viral DNA in cell cultures. Wavelength Maxima: Excitation 358nm, Emission 461nm

Formel: C₁₆H₁₅N₅·2HCl
Molecular Weight: 350,25 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00012681
CAS-Nummer: 28718-90-3
EINECS: 249-186-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0754f86117a82d55cf0db2e5171258993af018f4&vid=540aec82e4a46e27df6303e45706a9d41c7238c1&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR