Acridine orange (chlorid salt) 95 HPLC_ASSAY_METHOD, ultrapure

Lieferant: ENZO LIFE SCIENCES

Synonyme: AO, C.I. 46005, 3,6-Bis(dimethylamino)acridine hydrochloride, Basic Orange 14, N,N,N’,N’-Tetramethyl-3,6-acridinediamine hydrochloride, 3,6-Acridinediamine, Acridine orange hydrochloride

ENZ52405
ENZOENZ52405EA 106 EUR
ENZOENZ52405
Acridine orange (chlorid salt) 95 HPLC_ASSAY_METHOD, ultrapure
Acridinorange (Chloridsalz)

DNA dye


Acridine orange is a nucleic acid selective fluorescent cationic dye useful for cell cycle determination. It is cell-permeable, and interacts with DNA and RNA by intercalation or electrostatic attractions. When bound to DNA, it is very similar spectrally to fluorescein, with an excitation maximum at 502 nm and an emission maximum at 525 nm (green). When it associates with RNA, the excitation maximum shifts to 460 nm (blue) and the emission maximum shifts to 650 nm (red). Acridine Orange is also commonly used to non-specifically stain acidic organelles, such as lysosomes The dye is often used in epifluorescence microscopy. Wavelength Maxima: Excitation 502 nm, Emission 525 nm.

Formel: C₁₇H₁₉N₃·HCl·xH₂O
Molecular Weight: 301.81 g/mol (anhy)
Lagertemperatur: Gefrierschrank
MDL: MFCD00012660
CAS-Nummer: 65-61-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f2a1c4a6331682d613b970c45b0bbe9887bbcb2&vid=4a1852d76aab7b85e22ad5d170606c9eac900a9b&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR