FITC (Fluorescein-5-isothiocyanat, Fluoresceinisothiocyanat Isomer I) ≥95% (durch HPLC), ultrarein

Lieferant: ENZO LIFE SCIENCES

ENZ52451
ENZOENZ52451EA 325 EUR
ENZOENZ52451
FITC (Fluorescein-5-isothiocyanat, Fluoresceinisothiocyanat Isomer I) ≥95% (durch HPLC), ultrarein
FITC (Fluorescein-5-isothiocyanat, Fluoresceinisothiocyanat Isomer I)

Amine reactive dye


Fluorescein isothiocyanate (FITC) is an amine-reactive fluorescein derivative that yields conjugates with superior stability. Dye synthesis generally results in two isomers that differ slightly in protein binding and may elute differently under chromatographic conditions, complicating analysis in certain situations. The 5-isomer of FITC is the most widely used isoform and has a multitude of applications beyond protein peptide and nucleotide labeling. For example, it is commonly employed as a donor for FRET (Fluorescence Resonance Energy Transfer) based assay systems. An acceptor molecule, such as TRITC, is recommended in such FRET applications. Wavelength Maxima: Excitation 494 nm, Emission 520 nm.

Formel: C₂₁H₁₁NO₅S
Lagertemperatur: Gefrierschrank
MDL: MFCD00005063
CAS-Nummer: 3326-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ae27a4a94b2cc63b2932d34ca5a084149f4c8cf&vid=bf5a6e4c6228c40203023d8bf14622016445bba3&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR