5(6)-Tetramethylrhodamine isothiocyanate [5(6)-TRITC] ≥95% (durch HPLC), ultrarein
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Tetramethylrhodamine-5(6)-isothiocyanate, 5(6)-TRITC, MRITC, TRITC
Amine reactive dye
5(6)-TRITC is an amino-reactive labeling reagent that is widely used in preparing bioconjugates of proteins and nucleic acids. The resultant conjugates have similar spectral properties to those prepared from 5(6)-TAMRA, SE with improved stability. TRITC is widely used and has a multitude of applications beyond protein, peptide and nucleotide labeling For example, it can serve as an acceptor for FRET (Fluorescence Resonance Energy Transfer) based detection systems. FITC is recommended as a donor molecule in these FRET-based applications. Wavelength Maxima: Excitation 543 nm, Emission 571 nm.
Formel:
C₂₅H₂₁N₃O₃S Molecular Weight: 443,52 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00063517 CAS-Nummer: 95197-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...