Nilrot ≥95% (durch HPLC), ultrarein

Lieferant: ENZO LIFE SCIENCES

Synonyme: 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one

ENZ52551
ENZOENZ52551EA 165 EUR
ENZOENZ52551
Nilrot ≥95% (durch HPLC), ultrarein
Nilrot

Lipid dye


Nile red is a lipophilic probe that becomes intensely fluorescent in a hydrophobic environment, while it has minimal fluorescence in aqueous media. It is an excellent vital stain for the detection of intracellular lipid droplets by fluorescence microscopy, in cells such as adipocyes. Flow cytometry methods using a single staining with Nile red and double staining with Nile red and anti-CD3 monoclonal antibody have been employed to monitor peripheral leukocyte and lymphocyte phospholipidosis. Nile Red has also been used for rapid staining of proteins in SDS-polyacrylamide gel electrophoresis. Wavelength Maxima: Excitation 552 nm, Emission 636 nm.

Formel: C₂₀H₁₈N₂O₂
Molecular Weight: 318,38 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00011639
CAS-Nummer: 7385-67-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d56a58bd298dabc56b539f61b18bcb8db9db1d6&vid=3c793a59ad5bddd0cb12a17b903fb4aa197d05e4&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR