Fluorescent calcium indicator, GFP-CERTIFIED® FLUOFORTE®
Lieferant: ENZO LIFE SCIENCES
ENZ525550250
ENZOENZ525550250EA
410
EUR
ENZOENZ525550250
Fluorescent calcium indicator, GFP-CERTIFIED® FLUOFORTE®
Tests
Zelltests
GFP-CERTIFIED® FLUOFORTE® is a fluorescent calcium indicator relative to other red fluorescent dyes.
Can be used for the detection of intracellular calcium mobilisation across a broad spectrum of biological targets.
Abs/Em.: 524/ 551 nm
Bitte beachten Esterase hydrolysis is not required for live cell analysis via flow cytometry or fluorescence microscopy.