Green AP Chromogen/Substrate
Lieferant: ENZO LIFE SCIENCES
ENZACC1300030
ENZOENZACC1300030EA
321
EUR
ENZOENZACC1300030
Green AP Chromogen/Substrate
Proteine und Peptide
HIGHDEF® Green AP Chromogen/Substrate is intended to be used in conjunction with alkaline phosphatase (AP)-based immunostaining or in situ hybridisation systems.