Guanylylimidodiphosphate, trisodium salt ≥95% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: Gpp(NH)p, Guanosine 5′-[β,γ-imido]triphosphate trisodium salt, GMP-PNP, β:γ-Imidoguanosine 5′-triphosphate trisodium salt hydrate

ENZCHM1000005
ENZOENZCHM1000005EA 305 EUR
ENZOENZCHM1000005
Guanylylimidodiphosphate, trisodium salt ≥95% (durch HPLC)
Guanylylimidodiphosphate, trisodium salt

ADP-ribosylation factor activator.


A nonhydrolyzable GTP analog. Gpp(NH)p is the preferred GTP analog for activation of ADP-ribosylation factor (ARF).


Soluble in water (10 mg/ml).

Formel: C₁₀H₁₄N₆Na₃O₁₃P₃
Molecular Weight: 588,14 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 148892-91-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=593fce069b712c6476a404506f7f1b76327b6d0b&vid=808273e48ec28ed6b05dc08313f356318b461277&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥95% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR