Virginiamycin M1 ≥95% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Staphylomycin, Mikamycin A
ENZCHM1050005
ENZOENZCHM1050005EA
627
EUR
ENZOENZCHM1050005
Virginiamycin M1 ≥95% (durch HPLC)
Virginiamycin M1
Streptogramin antibiotic
Isolated from Streptomyces virginiae.
Antibiotic that inhibits bacterial protein synthesis.
Formel:
C₂₈H₃₅N₃O₇ Molecular Weight: 525,6 g/mol |
CAS-Nummer:
21411-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...