Virginiamycin S1 ≥99% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

ENZCHM1060005
ENZOENZCHM1060005EA 502 EUR
ENZOENZCHM1060005
Virginiamycin S1 ≥99% (durch HPLC)
Virginiamycin S1

Streptogramin antibiotic


Isolated from Streptomyces virginiae.

Antibiotic that inhibits bacterial protein synthesis.

Formel: C₄₃H₄₉N₇O₁₀
CAS-Nummer: 23152-29-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8187da72a5433918ef9d9ebeec83e4556270fc09&vid=2dcd287d000f6ec9fead156d23df9d5868355fa9&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR