PDGF receptor tyrosine kinase inhibitor IV ≥99% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
JNJ-10198409
ENZCHM1110005
ENZOENZCHM1110005EA
127
EUR
ENZOENZCHM1110005
PDGF receptor tyrosine kinase inhibitor IV ≥99% (durch HPLC)
PDGF receptor tyrosine kinase inhibitor IV
Cell-permeable PDGFR inhibitor with antiproliferative properties.
A cell-permeable compound acting as an ATP-competitive and reversible inhibitor of PDGFR. PDGF receptor tyrosine kinase inhibitor IV displays potent antiproliferative properties in several human tumor cell lines.
Formel:
C₁₈H₁₆FN₃O₂ Molecular Weight: 325,34 g/mol |
CAS-Nummer:
627518-40-5 |