SecinH3 ≥99% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: N-(4-(5-(Benzo[d][1,3]dioxol-5-yl)-3-methoxy-1H-1,2,4-triazol-1-yl)phenyl)-2-(phenylthio)acetamide

ENZCHM1140005
ENZOENZCHM1140005EA 172 EUR
ENZOENZCHM1140005
SecinH3 ≥99% (durch HPLC)
SecinH3

Cell-permeable cytohesin family small GEF antagonist.


A Sec7 domain-binding selective antagonist against cytohesin family small GEFs. SecinH3 inhibits large GEFs only at much higher concentrations and is shown to effectively inhibit insulin signaling.

Formel: C₂₄H₂₀N₄O₄S
Molecular Weight: 460.5 g/mol
MDL: MFCD10687099
CAS-Nummer: 853625-60-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d116e2386956ba3cc80ae1018a6e226da43bff7f&vid=76258a7c75dffa05c843c7eac4e5b92a16ed65f8&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR