FABP4 inhibitor ≥99% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
BMS-309403, 2-((2'-(5-Ethyl-3,4-diphenyl-1H-pyrazol-1-yl)-[1,1'-biphenyl]-3-yl)oxy)acetic acid
ENZCHM1180005
ENZOENZCHM1180005EA
102
EUR
ENZOENZCHM1180005
FABP4 inhibitor ≥99% (durch HPLC)
FABP4 inhibitor
A cell-permeable, potent, and selective inhibitor of FABP4.
Formel:
C₃₁H₂₆N₂O₃ Molecular Weight: 474,5500 g/mol |
MDL:
MFCD09991687 CAS-Nummer: 300657-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...