MIF antagonist, ISO-1 ≥97% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Methyl 2-(3-(4-hydroxyphenyl)-4,5-dihydroisoxazol-5-yl)acetate, ISO-1
ENZCHM1210005
ENZOENZCHM1210005EA
131
EUR
ENZOENZCHM1210005
MIF antagonist, ISO-1 ≥97% (durch HPLC)
MIF antagonist, ISO-1
Cell permeable MIF antagonist with anti-inflammatory properties.
Formel:
C₁₂H₁₃NO₄ Molecular Weight: 235,2 g/mol |
MDL:
MFCD22376813 CAS-Nummer: 478336-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...