Simvastatin sodium salt ≥97% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
ENZCHM1420005
ENZCHM1420010
ENZOENZCHM1420005EA
172
EUR
ENZOENZCHM1420005
ENZOENZCHM1420010
Simvastatin sodium salt ≥97% (durch HPLC)
Simvastatin sodium salt
Competitive inhibitor of HMG-CoA reductase. Suppresses TNF-induced NF-κB activation.
Simvastatin is marketed under the trade name Zocor™ and is often prescribed in combination with ezetimibe (Zetia) to treat dyslipidemia. This drug combination is known commercially as Vytorin or Inegy. Simvastatin also suppresses TNF-induced NF-κB activation, which potentiates apoptosis in human myeloid leukemia cells and thus, may be useful in treating cancer.
Molecular Weight:
458.6 g/mol |
CAS-Nummer:
101314-97-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...