NSC697923 ≥99% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 2-(4-Methylphenyl)sulphonyl-5-nitrofuran, 2-Nitro-5-tosylfuran, NSC-697923

ENZCHM1430005
ENZOENZCHM1430005EA 70.4 EUR
ENZOENZCHM1430005
NSC697923 ≥99% (durch HPLC)
NSC697923

Cell-permeable and selective inhibitor of the ubiquitin-conjugating (E2) enzyme Ubc13-Uev1A.

Formel: C₁₁H₉NO₅S
Molecular Weight: 267,26 g/mol
MDL: MFCD01925132
CAS-Nummer: 343351-67-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e279dcda0ab18b05c654c3332e8320f50777e32&vid=ae457ed35ab81111ef5b21cdd201cd6e6f6951b0&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR