Hydrogenchlorid --N-(3-ethinylphenyl)-6,7-bis(2-methoxyethoxy)-4-chinazolinamin ≥95% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Erlotinib Hydrochloride

ENZCHM1570050
ENZOENZCHM1570050EA 227 EUR
ENZOENZCHM1570050
Hydrogenchlorid --N-(3-ethinylphenyl)-6,7-bis(2-methoxyethoxy)-4-chinazolinamin ≥95% (durch HPLC)
N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride

EGFR tyrosine kinase auto-phosphorylation inhibitor


Erlotinib inhibits EGFR tyrosine kinase auto-phosphorylation. Studies in cell lines and enzyme assays have shown that erlotinib inhibits EGFR at concentrations significantly lower than those needed to inhibit c-src and v-abl. It has an IC50 of 2 nM against EGFR. It is >1000-fold more sensitive for EGFR than for c-Src or v-Abl.

Formel: C₂₂H₂₄ClN₃O₄
Molecular Weight: 429,901 g/mol
MDL: MFCD07781272
CAS-Nummer: 183319-69-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91875cea23b4a5a7a5a23263d37c4fbb0b1118f5&vid=f8152b32b0bf33cf8acac282e590269a177037bd&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR